n-Dodecyl-β-D-maltoside
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: DDM
ALX500007G001 ALX500007G005 ALX500007G025
ENZOALX500007G001EA 178 EUR
ENZOALX500007G001 ENZOALX500007G005 ENZOALX500007G025
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside
Formule: C₂₄H₄₆O₁₁
MW: 510.6 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00043012
CAS nummer: 69227-93-6