Luminol (3-Aminophthalhydrazide) ≥97% (by HPLC)
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: NSC 5064
ALX610002G001 ALX610002G005
ENZOALX610002G001EA 49.5 EUR
ENZOALX610002G001 ENZOALX610002G005
Luminol (3-Aminophthalhydrazide) ≥97% (by HPLC)
Luminol (3-Aminophthalhydrazide)
Formule: C₈H₇N₃O₂
MW: 177.16 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00006890
CAS nummer: 521-31-3
EINECS: 208-309-4