Patton-Reeders reagent (Calconcarboxylic acid), (max. 10% H₂O)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B23459.09
B23459.18
B23459.09EA
59.9
EUR
B23459.09
B23459.18
Patton-Reeders reagent (Calconcarboxylic acid), (max. 10% H₂O)
Patton-Reeders reagent (Calconcarboxylic acid)
CAS No.: 3737-95-9
Molecular Formula: C21H14N2O7S
Formula Weight: 438.42
Melting Point: ca 300° dec.
Beilstein Reference: 727492
MDL No.: MFCD00004078
Formule:
C₂₁H₁₄N₂O₇S Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004078 CAS nummer: 3737-95-9 EINECS: 223-117-0 |