Pvnhars

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Rosin, gum, Rosin, Gum rosin

9051.1 9051.2
ROTH9051.1EA 0 EUR
ROTH9051.1 ROTH9051.2
Pvnhars
Pvnhars
Smeltpunt: 75…81 °C
Dichtheid: 1.1 g/cm³ (20 °C)
Vlampunt: 188
MDL Nummer: MFCD00132205
CAS nummer: 8050-09-7
EINECS: 232-475-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR