Fenolrood natriumzout 0,1% in ethanol (20%) Reag. Ph. Eur. 1063601
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
87888.180EA
44.3
EUR
87888.180DP
87888.180
87888.180_T
Fenolrood natriumzout 0,1% in ethanol (20%) Reag. Ph. Eur. 1063601
Fenolrood natriumzout
Bestelinformatie: Normally manufactured to order. Please check with customer services.
Formule:
C₁₉H₁₃NaO₅S MW: 376,35 g/mol |
MDL Nummer:
MFCD00066901 CAS nummer: 34487-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...