Pyridinium chlorochromate
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: PCC
SB01867.1 SB01867.50
MAYBSB01867.1EA 0 EUR
MAYBSB01867.1 MAYBSB01867.50
Pyridinium chlorochromate
Pyridinium chlorochromate
Formule: C₅H₆ClCrNO₃
MW: 215.56 g/mol
Smeltpunt: 205
MDL Nummer: MFCD00013106
CAS nummer: 26299-14-9
EINECS: 247-595-5
UN: 1479
ADR: 5.1 II
Merck Index: 13,08064