Saffron, Sigma-Aldrich®
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: C.I. 75100
M-Clarity™ quality level = MQ200
S8381-1G S8381-5G
S8381-1GEA 129 EUR
S8381-1G S8381-5G
Saffron, Sigma-Aldrich®
Saffron
MW: 976.96 g/mol
CAS nummer: 84604-17-1
EINECS: 283-295-0