Thymol blue sodium salt 0,04% (w/v) in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bis[5-methyl-2-(1-methylethyl)phenolate] S,S-dioxide, Sodium 4-(3-(4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-isopropyl-5-methylphenolate, Thymol blue water soluble, Thymol Blue sodium
38692.AE 38692.AP
38692.AEEA 23.9 EUR
38692.AE 38692.AP
Thymol blue sodium salt 0,04% (w/v) in aqueous solution
Thymol blue sodium salt
CAS No.: 62625-21-2 Molecular Formula: C27H29NaO5S Formula Weight: 488.58 Physical Form: Liquid Density: 0.979 Odour: Odourless MDL No.: MFCD00151093
Formule: C₂₇H₂₉NaO₅S
MW: 488.58 g/mol
Smeltpunt: 283
Dichtheid: 0.979
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00151093
CAS nummer: 62625-21-2
EINECS: 200-973-3