Naples Women's Polo Shirts
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B209BKRL B209BKRM B209BKRS B209BKRXL B209BKRXS B209BKRXXL
PRTWB209BKRLEA 13.1 EUR
PRTWB209BKRL PRTWB209BKRM PRTWB209BKRS PRTWB209BKRXL PRTWB209BKRXS PRTWB209BKRXXL
Naples Women's Polo Shirts
Werkkleding
Beautifully fitted and ideal for personalisation, this essential women's style meets all workwear needs.
  • Ideal for corporate wear and custom finishes
  • Button placket opening
  • Rib collar and cuffs
  • Twin-stitched seams for extra durability
  • 40+ UPF rated fabric to block 98% of UV rays
Made using pique knit polycotton fabric which is soft to touch and comfortable to wear. Recommended for dry daily wear.
Order Now
Specificaties
Kledingstuk toepassing General Purpose General Purpose General Purpose General Purpose General Purpose
Environmentally Preferable
Risk area
Geslacht Voor vrouwen Voor vrouwen Voor vrouwen Voor vrouwen Voor vrouwen
Sleeve or leg length Short Short Short Short Short
Stof 65% polyester, 35% cotton 65% polyester, 35% cotton 65% polyester, 35% cotton 65% polyester, 35% cotton 65% polyester, 35% cotton
Gewicht 0,248 g/m² 0,25 g/m² 0,302 g/m² 0,2 g/m² 0,303 g/m²