Vanquish Rain Trousers
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S556BKRL S556BKRM S556BKRS S556BKRXL S556BKRXXL
PRTWS556BKRLEA 39.5 EUR
PRTWS556BKRL PRTWS556BKRM PRTWS556BKRS PRTWS556BKRXL PRTWS556BKRXXL
Vanquish Rain Trousers
Werkkleding
A complimentary trouser style to tie into the PWR rainwear collection. The vanquish trouser uses the highly waterproof and breathable Portwest extreme fabric with taped seams to ensure full protection.
  • Abrasion-resistant oxford polyester
  • 2 side zip pockets for secure storage
  • Elastic waist with toggle for a secure and comfortable fit
  • Side zip leg opening for easy access
  • Double-layer reinforced knee protection for extra strength and durability
  • Taped seams to provide additional protection
  • Fully elasticated waistband for ultimate wearer comfort
Durable, breathable, windproof and water-resistant fabric. The extremely water resistant eco-conscious fabric finish beads water away from the fabric surface. A zipped side leg allows for easy access on and off while wearing boots. A fully elasticated, adjustable waistband and articulated legs ensure an excellent fit.
Recommended for wet weather wear. Black finish.
Certificaten: EN 343 Class 3:3 X WP 10,000 mm, MVP 15,000 g/M²/24 hr. CE certified.