Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (APC (Allophycocyanin)/Cy7®)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

104411D0 104411D1 104411D2
AATB104411D0EA 319 EUR
AATB104411D0 AATB104411D1 AATB104411D2
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (APC (Allophycocyanin)/Cy7®)
Antilichamen
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Specificaties

antigeen symbool CD44
Environmentally Preferable
Type antilichaam Primair,CD44 antigen
Type antilichaam Primair,CD44 antigen
Clonality Monoklonaal
Conjugatie APC (Allophycocyanin)/Cy7®
Clone IM7
Reactivity Human
Host Rat
Isotype
ImmunoFluorescentie Yes
Flowcytometrie Yes
Purification Affinity chromatography
Kruisadsorptie No
Opslagtemperatuur 2...8 °C with minimised light exposure. Do not freeze.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR