Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 810)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

100320O0 100320O1
AATB100320O0EA 371 EUR
AATB100320O0 AATB100320O1
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 810)
Antilichamen
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kDa member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: iFluor® 810
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR