Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (mFluor™ Blue 570)
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100320T0 100320T1
AATB100320T0EA 371 EUR
AATB100320T0 AATB100320T1
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (mFluor™ Blue 570)
Antilichamen
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kDa member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Opgelet: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: mFluor™ Blue 570
Epitope:
Host: Mouse
Isotype:
Reactivity: Human