Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (Pacific Orange)
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
101921K0 101921K1
AATB101921K0EA 371 EUR
AATB101921K0 AATB101921K1
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (Pacific Orange)
Antilichamen
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Opgelet: For research use only.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: Pacific Orange
Epitope:
Host: Mouse
Isotype:
Reactivity: Human