Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (Pacific Orange)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

103401K0 103401K1
AATB103401K0EA 371 EUR
AATB103401K0 AATB103401K1
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (Pacific Orange)
Antilichamen
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: Pacific Orange
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR