Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PE (Phycoerythrin)/iFluor® 700)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

100411X0 100411X1 100411X2
AATB100411X0EA 319 EUR
AATB100411X0 AATB100411X1 AATB100411X2
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PE (Phycoerythrin)/iFluor® 700)
Antilichamen
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: PE (Phycoerythrin)/iFluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR