Assistent Brushes for Bottles
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41279010 91120000015 91231200030 91231200045 91231200060 91231200090
HECH41279010EA 8.4 EUR
HECH41279010 HECH91120000015 HECH91231200030 HECH91231200045 HECH91231200060 HECH91231200090
Assistent Brushes for Bottles
Borstels
An assortment of brushes, available with nylon or natural bristles, designed for use with laboratory bottles and flasks.
  • For steep-bottom, round-shoulder, and threaded bottles and flasks
Brushes feature a head bundle for better cleaning of the bottoms of bottles and flasks.