Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 532)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

11610070 11610071
AATB11610070EA 371 EUR
AATB11610070 AATB11610071
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 532)
Antilichamen
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 532
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR