Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 560)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

102800A0 102800A1
AATB102800A0EA 371 EUR
AATB102800A0 AATB102800A1
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 560)
Antilichamen
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: iFluor® 560
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR