Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 568)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

116100B0 116100B1
AATB116100B0EA 371 EUR
AATB116100B0 AATB116100B1
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 568)
Antilichamen
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 568
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR