Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 670)
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
102800H0 102800H1
AATB102800H0EA 371 EUR
AATB102800H0 AATB102800H1
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 670)
Antilichamen
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Opgelet: For research use only.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: iFluor® 670
Epitope:
Host: Mouse
Isotype:
Reactivity: Human