Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Violet 450)
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100330Z0 100330Z1
AATB100330Z0EA 371 EUR
AATB100330Z0 AATB100330Z1
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Violet 450)
Antilichamen
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Opgelet: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: mFluor™ Violet 450
Epitope:
Host: Mouse
Isotype:
Reactivity: Human