Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ Red 780)
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
104410W0 104410W1
AATB104410W0EA 371 EUR
AATB104410W0 AATB104410W1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ Red 780)
Antilichamen
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Opgelet: For research use only.
Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: mFluor™ Red 780
Epitope:
Host: Rat
Isotype:
Reactivity: Human