Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (mFluor™ Violet 510)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

10051110 10051111
AATB10051110EA 371 EUR
AATB10051110 AATB10051111
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (mFluor™ Violet 510)
Antilichamen
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD5 antigen
Clonality: Monoclonal
Clone: L17F12
Conjugation: mFluor™ Violet 510
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR