Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® 555)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

10141090 10141091
AATB10141090EA 371 EUR
AATB10141090 AATB10141091
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® 555)
Antilichamen
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: iFluor® 555
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Specificaties

antigeen symbool CD14
Type antilichaam Primair,CD14 antigen
Environmentally Preferable
Type antilichaam Primair,CD14 antigen
Clonality Monoklonaal
Conjugatie iFluor® 555
Clone 61D3
Reactivity Human
Host Mouse
Isotype
ImmunoFluorescentie Yes
Flowcytometrie Yes
Purification Affinity chromatography
Kruisadsorptie No
Opslagtemperatuur 2...8 °C with minimized light exposure. Do not freeze.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR