Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® 810)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

101410O0 101410O1
AATB101410O0EA 371 EUR
AATB101410O0 AATB101410O1
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (iFluor® 810)
Antilichamen
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: iFluor® 810
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR