Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (mFluor™ Violet 540)
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10052120 10052121
AATB10052120EA 371 EUR
AATB10052120 AATB10052121
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (mFluor™ Violet 540)
Antilichamen
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Opgelet: For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: mFluor™ Violet 540
Epitope:
Host: Mouse
Isotype:
Reactivity: Human