Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PB (Pacific Blue))
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
116101J0 116101J1
AATB116101J0EA 371 EUR
AATB116101J0 AATB116101J1
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PB (Pacific Blue))
Antilichamen
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Opgelet: For research use only.
Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human