Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (Pacific Orange)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

100331K0 100331K1
AATB100331K0EA 371 EUR
AATB100331K0 AATB100331K1
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (Pacific Orange)
Antilichamen
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Opgelet: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: Pacific Orange
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR