Pipette adapters
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5833000 5833001
MARI5833000EA 95.7 EUR
MARI5833000 MARI5833001
Pipette adapters
Pipetten
Adapters made of PS. Suitable for tubings for blood diluting pipettes and haemoglobin pipettes.
Not registered for in vitro diagnostic use within the EU.