BizTex® Microporous Coveralls, Type 3/4/5/6
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ST70YERL ST70YERM ST70YERXL ST70YERXXL
PRTWST70YERLEA 897 EUR
PRTWST70YERL PRTWST70YERM PRTWST70YERXL PRTWST70YERXXL
BizTex® Microporous Coveralls, Type 3/4/5/6
Werkkleding
This coverall offers exceptional mechanical strength, as well as liquid and particle protection.
  • Excellent liquid and particulate barrier protection
  • Double storm flap to protect against the elements
  • Thumb loop
  • Elasticated cuffs for a secure fit
  • Ergonomically shaped hood for a perfect fit
  • Retail bag aids presentation for retail sales
The durable microporous polypropylene laminate provides an excellent barrier to chemical spray and infective agents. The fabric's physical strength and flexibility ensures protection and comfort in the harshest environments.
Certificaten: EN 14605 Type 3B. EN 14605 Type 4B. EN ISO 13982 Type 5B. EN 13034 Type 6B. EN 1073-2. EN 1149 -5. EN 14126. CE-CAT III.