ESD Safety Ankle Boots, Lacing Sytem, Ian XXTP Pro Boa® Mid S3 S
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
761331/35 761331/36 761331/37 761331/38 761331/39 761331/40 761331/41 761331/42 761331/43 761331/44 761331/45 761331/46 761331/47 761331/48 761331/49
ELTN761331/35EA 121.9 EUR
ELTN761331/35 ELTN761331/36 ELTN761331/37 ELTN761331/38 ELTN761331/39 ELTN761331/40 ELTN761331/41 ELTN761331/42 ELTN761331/43 ELTN761331/44 ELTN761331/45 ELTN761331/46 ELTN761331/47 ELTN761331/48 ELTN761331/49
ESD Safety Ankle Boots, Lacing Sytem, Ian XXTP Pro Boa® Mid S3 S
Schoenen en laarzen
ESD safety ankle boots made from hydrophobised microfibre/hydrophobised textile upper and a TPU/PU sole (Wellmaxx Trainers Power).
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Boa® fit lacing system
  • Non-metallic, penetration-resistant midsole
  • Composite toe cap
  • TPU scuff cap
Suitable for areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Blue
Certificaten: EN ISO 20345 S3S FO/SR/SC, form B.