ESD Safety Shoes, Lace-up, Low, Jori jo_Champ Low S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12421/36 12421/37 12421/38 12421/39 12421/46 12421/40 12421/41 12421/42 12421/43 12421/44 12421/45 12421/47 12421/48
ELTN12421/36EA 49.9 EUR
ELTN12421/36 ELTN12421/37 ELTN12421/38 ELTN12421/39 ELTN12421/46 ELTN12421/40 ELTN12421/41 ELTN12421/42 ELTN12421/43 ELTN12421/44 ELTN12421/45 ELTN12421/47 ELTN12421/48
ESD Safety Shoes, Lace-up, Low, Jori jo_Champ Low S3
Schoenen en laarzen
ESD safety shoe made from cowhide leather upper and a PU/PU sole (jo_Explore).
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole jo_TITAN ESD
  • Soft fleece ESD insole
  • Steel midsole
  • Steel toe cap
  • PU scuff cap
Suitable for indoors and outdoors, areas where exposure to moisture is expected (S2), areas where there is a risk of penetration from pointed and sharp objects (S3), areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black
Certificaten: EN ISO 20345 S3 FO/LG/SR, form A.