ESD Occupational Shoes, Lace-up, Ronan XXF GTX O2 WR CI Low
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
972620/45 972620/46 972620/47 972620/48 972620/35 972620/36 972620/37 972620/38 972620/39 972620/40 972620/41 972620/42 972620/43 972620/44
ELTN972620/45EA 119.9 EUR
ELTN972620/45 ELTN972620/46 ELTN972620/47 ELTN972620/48 ELTN972620/35 ELTN972620/36 ELTN972620/37 ELTN972620/38 ELTN972620/39 ELTN972620/40 ELTN972620/41 ELTN972620/42 ELTN972620/43 ELTN972620/44
ESD Occupational Shoes, Lace-up, Ronan XXF GTX O2 WR CI Low
Schoenen en laarzen
ESD work shoe made from hydrophobized nubuck leather and a PU/PU sole (Wellmaxx Flex).
  • Full, padded bellows tongue
  • All weather Gore-Tex performance membrane
  • Full-length inlay sole ESD PRO black
  • Soft fleece ESD insole
Suitable for indoors and outdoors, areas where exposure to moisture is expected (O2), areas where there is no risk of falling heavy objects, areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Dark grey
Certificaten: EN ISO 20347 O2 WR/FO/SR/CI, form A.