ESD Safety Ankle Boots, Lace-Up, Jo_Champ Mid S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
16421/36 16421/37 16421/38 16421/39 16421/40 16421/41 16421/42 16421/43 16421/44 16421/45 16421/46 16421/47 16421/48
ELTN16421/36EA 53.9 EUR
ELTN16421/36 ELTN16421/37 ELTN16421/38 ELTN16421/39 ELTN16421/40 ELTN16421/41 ELTN16421/42 ELTN16421/43 ELTN16421/44 ELTN16421/45 ELTN16421/46 ELTN16421/47 ELTN16421/48
ESD Safety Ankle Boots, Lace-Up, Jo_Champ Mid S3
Schoenen en laarzen
ESD safety ankle boot made from cowhide leather upper and a PU/PU sole (jo_EXPLORE).
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Soft fleece ESD insole
  • Full-length inlay sole (jo_TITAN ESD)
  • Steel midsole
  • Steel toe cap
  • PU scuff cap
Suitable for indoors and outdoors, areas where exposure to moisture is expected (S2), where there is a risk of penetration from pointed and sharp objects (S3) and/or where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black
Certificaten: EN ISO 20345 S3 FO/LG/SC/SR, form B.