ESD Safety Ankle Boots, Lace-Up, Leonardo XSG Mid S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
768711/36 768711/37 768711/38 768711/39 768711/40 768711/41 768711/42 768711/43 768711/44 768711/45 768711/46 768711/47 768711/48
ELTN768711/36EA 80.9 EUR
ELTN768711/36 ELTN768711/37 ELTN768711/38 ELTN768711/39 ELTN768711/40 ELTN768711/41 ELTN768711/42 ELTN768711/43 ELTN768711/44 ELTN768711/45 ELTN768711/46 ELTN768711/47 ELTN768711/48
ESD Safety Ankle Boots, Lace-Up, Leonardo XSG Mid S3
Schoenen en laarzen
ESD safety ankle boot made from cowhide leather upper and a PU/PU sole (Wellmaxx Safety Grip).
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ARTISAN ESD
  • Non-metallic, penetration-resistant midsole
  • PU toe protection and steel toe cap
This boots are suitable for use both indoors and outdoors, in areas where exposure to moisture is expected (S2), where there is a risk of penetration from pointed and sharp objects (S3), and where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black/blue
Certificaten: EN ISO 20345 S3 SRC, form B.