ESD Safety Ankle Boots, Maddox Boa® Mid S3 S
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
769151/35 769151/36 769151/37 769151/38 769151/39 769151/40 769151/41 769151/42 769151/43 769151/44 769151/45 769151/46 769151/47 769151/48
ELTN769151/35EA 111.9 EUR
ELTN769151/35 ELTN769151/36 ELTN769151/37 ELTN769151/38 ELTN769151/39 ELTN769151/40 ELTN769151/41 ELTN769151/42 ELTN769151/43 ELTN769151/44 ELTN769151/45 ELTN769151/46 ELTN769151/47 ELTN769151/48
ESD Safety Ankle Boots, Maddox Boa® Mid S3 S
Schoenen en laarzen
Made of hydrophobised microfibre/Hydrophobised textile material.
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole SPORTIVE ESD black
  • Non-metallic, penetration-resistant midsole
  • TPU/PU sole MAXXIMO
  • Steel toe cap
  • BOA® Fit System
  • Leather-free equipment, Sole core made of Infinergy® by BASF
Certificaten: EN ISO 20345:2022 S3S FO/SR, form B.