ESD Safety Ankle Boots, Lacing Sytem, Stefano XXSG Boa® Mid S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
768741/41 768741/36 768741/37 768741/38 768741/39 768741/40 768741/42 768741/43 768741/44 768741/45 768741/46 768741/47 768741/48
ELTN768741/41EA 124.9 EUR
ELTN768741/41 ELTN768741/36 ELTN768741/37 ELTN768741/38 ELTN768741/39 ELTN768741/40 ELTN768741/42 ELTN768741/43 ELTN768741/44 ELTN768741/45 ELTN768741/46 ELTN768741/47 ELTN768741/48
ESD Safety Ankle Boots, Lacing Sytem, Stefano XXSG Boa® Mid S3
Schoenen en laarzen
ESD safety ankle boot made from cowhide leather upper and a PU/PU sole (Wellmaxx Safety Grip).
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ARTISAN ESD
  • Boa® fit lacing system
  • Non-metallic, penetration-resistant midsole
  • Steel toe cap
  • PU toe protection
Suitable for indoors and outdoors, areas where exposure to moisture is expected (S2), areas where there is a risk of penetration from pointed and sharp objects (S3), areas where there is a risk of electrostatic discharge (ESDS/ESD), Workplaces on hard undergrounds: The revolutionary Infinergy® sole core cushions impacts and provides for a rebound effect when the compressive impulse subsides - for more energy in every step.
Colour: Black/blue
Certificaten: EN ISO 20345 S3 SRC, form B.