ESD Safety Shoes, Lacing System, Joran Boa® Low S3 L
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720861/39 720861/36 720861/37 720861/38 720861/40 720861/41 720861/42 720861/43 720861/44 720861/45 720861/46 720861/47 720861/48
ELTN720861/39EA 109.9 EUR
ELTN720861/39 ELTN720861/36 ELTN720861/37 ELTN720861/38 ELTN720861/40 ELTN720861/41 ELTN720861/42 ELTN720861/43 ELTN720861/44 ELTN720861/45 ELTN720861/46 ELTN720861/47 ELTN720861/48
ESD Safety Shoes, Lacing System, Joran Boa® Low S3 L
Schoenen en laarzen
ESD safety shoe made of hydrophobised microfibre and textile upper and a rubber/TPU/EVA sole (Transformers).
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole ESD
  • Boa® fit lacing system
  • Non-metallic, penetration-resistant midsole
  • Composite toe cap
  • Rubber scuff cap
Areas where there is a risk of electrostatic discharge (ESDS/ESD).
Certificaten: EN ISO 20345 S3L FO/HRO/SC/SR, form A.
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering ISO 14001