GelGreen® Agarose LE
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GelGreen®
41030-50GEA 0 EUR
41030-50G
GelGreen® Agarose LE
Gel voor elektroforese Agarose en acrylamide
Ultrapure molecular biology grade agarose pre-coated with GelGreen® Nucleic acid gel stain for convenient and safer preparation of precast gels.
  • Non-toxic GelGreen® DNA dye is already in the agarose
  • No need to handle concentrated dye solutions, for enhanced safety
  • Use with TAE or TBE for 0,8 to 2% gels
  • Low EEO, ultrapure molecular biology grade agarose
  • Compatible with blue light illuminators
GelGreen® Agarose LE is our ultra-pure molecular biology grade LE agarose pre-coated with GelGreen® Nucleic acid gel stain. There is no need to handle concentrated fluorescent dye while preparing your gel, for greater convenience and safety. For agarose gels from 0,8 to 2% in TAE or TBE.
GelGreen® is a sensitive, stable and environmentally safe green fluorescent nucleic acid dye specifically designed for gel staining. GelGreen® is far more sensitive than SYBR® Safe. Unlike SYBR® dyes, which are known to be unstable, GelGreen® is very stable, both hydrolytically and thermally. GelGreen® has UV absorption between 250 and 300 nm and a strong absorption peak centered around 500 nm.