Bacillol® 30 Sensitive Green Tissues
Leverancier: BODE CHEMIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9819430 9819430_1
BODE9819430EA 0 EUR
BODE9819430 BODE9819430_1
Bacillol® 30 Sensitive Green Tissues
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Desinfectiedoekjes
The ready-to-use Bacillol® 30 sensitive green tissues disinfectant wipes from the flowpack, soaked with Bacillol® 30 sensitive foam, wet the surfaces excellently and work quickly, even on sensitive surfaces.
  • 100% plastic-free wipes
  • Free of microplastics
  • Fast-acting with good cleaning properties
  • Particularly gentle on materials, also suitable for sensitive materials such as: polycarbonate (PC), acrylic glass (PMMA) and polysulfone
  • It can be used without gloves
  • Skin-friendly and allergy-friendly
Effectiveness Bactericidal, yeasticidal, tuberculocidal, mycobactericidal, limited virucidal (incl. HBV, HIV, HCV), limited virucidal PLUS.
Certificaten: ​​SGS Institut Fresenius**- and ECARF-certified.