Removable Blackout Labels for Thermal Transfer Printers
Leverancier: GA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AZA152NPC1-1WH AZA152NPC3-6WH AZA-67C1-1WH AZA-67C3-3WH
GAINAZA152NPC1-1WHEA 44.7 EUR
GAINAZA152NPC1-1WH GAINAZA152NPC3-6WH GAINAZA-67C1-1WH GAINAZA-67C3-3WH
Removable Blackout Labels for Thermal Transfer Printers
Etiketten
These removable labels have a unique opaque design that will obscure all pre-existing information. Can be used to re-identifying slides marked with unclear, erroneous, or incomplete information, but can also be removed or repositioned without leaving behind any residue.
  • Unique opaque design allows the labels to obscure all pre-existing information
  • Removable adhesive ensures easy removal without damaging information underneath or leaving adhesive residue
  • Ideal for relabeling microscope slides
  • Thermal transfer printable with all major barcode printers
Cover existing information with these easily removable blackout labels made to fit microscope slides. Ideal for re-identifying slides marked with unclear, erroneous, or incomplete information. These waterproof labels are made with a durable thermoplastic film and leave behind no residue when removed. Available in roll format and compatible for use with all major brands of thermal transfer printers.