Mono Pocket Samplers
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1430S-1000 1430S-1500 1430S-600
SAMP1430S-1000EA 726 EUR
SAMP1430S-1000 SAMP1430S-1500 SAMP1430S-600
Mono Pocket Samplers
Monsternemers Monsternemers voor vaste stoffen
The mono pocket sampler is designed for sampling free-flowing powders and granules from a single point.
  • Great for taking a single sample of free-flowing powders/granules
  • Available in a range of sizes
  • Can be easily stripped down for cleaning
  • Available from stock
  • Fast delivery
The mono pocket sampler is suitable for taking samples of free-flowing powders/granules from a single point. Available in various lengths, the GMP design ensures excellent durability for many applications.