Stopper, IN-Stopper
Leverancier: Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-2581EA 79.5 EUR
720-2581
Stopper, IN-Stopper
Stoppen PE/LDPE/HDPE stoppen
IN-Stopper is a male luer lock closing cone with injection port for intermittent injections through injection membrane. It closes female accesses and offers a needle access via an injection membrane, e.g. to an infusion solution and has got a smooth, hygienic outer surface.
  • Smooth, hygienic outer surface
  • Offers a needle access via a membrane, which is not made with Latex
  • IN-Stopper can be used with alcohol and lipids
  • IN-Stopper are non-toxic and non-pyrogenic
  • Easy and convenient in handling
  • Five insertions possible
The device can be used for all patients for which infusion therapy is prescribed. No gender or age-related limitations. IN-Stopper can be used for adults, pediatric and neonates.