Point Sampler
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1640A-1000 1640A-2000
SAMP1640A-1000EA 1710 EUR
SAMP1640A-1000 SAMP1640A-2000
Point Sampler
Monsternemers Monsternemers voor vaste stoffen
The point sampler is designed for large samples of powders.
  • Ideal for taking large volumes of powder from a single point
  • Perfect for both free-flowing and sticky powders
  • Easy to operate
  • Available from stock
  • Fast delivery
The point sampler is able to quickly take a single, large volume sample of both free-flowing or cohesive powders. The easy-to-use design means the scraper pushes the product into the sampler when the device is rotated.
Order Now
Specificaties
Environmentally Preferable