Silo Rocket Sampler
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1985A-1000 1985A-1500 1985A-2000
SAMP1985A-1000EA 2690 EUR
SAMP1985A-1000 SAMP1985A-1500 SAMP1985A-2000
Silo Rocket Sampler
Monsternemers Monsternemers voor vaste stoffen
The Silo rocket sampler is ideal for taking large bulk samples.
  • Suitable for both cohesive and free-flowing powders
  • Can be used for grains, granules, etc.
  • Fitted with a wide handle for extra leverage
  • Available from stock
  • Fast delivery
The Silo rocket sampler features a convenient design, meaning it can easily take samples of both free-flowing and cohesive powders. It is made from high-quality 316L stainless steel for durability.