Fluid Point Sampler
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5551S-600 5551S-1000 5551S-1500
SAMP5551S-600EA 704 EUR
SAMP5551S-600 SAMP5551S-1000 SAMP5551S-1500
Fluid Point Sampler
Monsternemers Monsternemers voor vloeistoffen
The fluid point sampler is designed for taking rapid samples of free-flowing liquids.
  • Ideal for sampling free-flowing liquids from most containers
  • Great for sampling pharmaceuticals, cosmetics, and foodstuffs
  • Easily cleaned
  • Available from stock
  • Fast delivery
The fluid point sampler is great for taking samples of free-flowing liquids from a range of containers, including drums. It is made from chemically resistant 316L stainless steel to provide excellent corrosion resistance.