Tight Head Drums
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A266-10 A266-20 A266-50 A266-100 A266-200
SAMPA266-10EA 499 EUR
SAMPA266-10 SAMPA266-20 SAMPA266-50 SAMPA266-100 SAMPA266-200
Tight Head Drums
Vaten, vaten, trommels
Tight head drums are perfect for use in food, chemical, pharmaceutical, and cosmetic manufacturing applications.
  • Made from high quality 316L stainless steel
  • Fitted with a 2" and ³/₄" bung
  • Comes with an EPDM seal
  • Available from stock
  • Fast delivery
High-quality stainless steel, closed top drums are ideal for use across several industries, including pharma and food. They area available in a wide range of sizes and have swaged sides for easier handling and added strength.