Emptying Rack
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SAMP1220R-1EA 633 EUR
SAMP1220R-1
Emptying Rack
Rekken Samplerrekken
Desiged for the convenient emptying of powder samples.
  • Enables the easy emptying of multi-level samplers
  • Made from 316L stainless steel
  • Supplied with 3 funnels and 3 bottles
  • Available from stock
  • Fast delivery
Emptying racks make the emptying of multi-level powder samplers extremely easy. Simply put the sampler on the rack and adjust the position of the funnels so they line up with the pockets. Open the sampler and the powder will fall into the sample bottles positioned underneath the funnels.
Note: for samplers over 1m please order 2 racks.