Stainless Screwtop Bottles
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A846-500 A846-1000
SAMPA846-500EA 457 EUR
SAMPA846-500 SAMPA846-1000
Stainless Screwtop Bottles
Flessen Metalen flessen
Sturdy and durable stainless screwtop bottles are designed for the most demanding applications.
  • Made from high-quality 316L stainless steel
  • Complete with a screw-on lid
  • Crevice-free and high-quality mirror polish finish
  • Ideal for the storage of powders and liquids
  • Available from stock
  • Fast delivery
This range of bottles are perfect for handling the most demanding powder/liquid storage applications. Made from high-quality 316L stainless steel, they come complete with a screw-on lid. The crevice-free, mirror polish finish ensures they are extremely easy to clean.